Gene Transcripts

Gene Symbol
TXN
Entrez Gene ID
7295
Gene Name
thioredoxin
Gene Aliases
TRDX, TRX, TRX1
Chromosome Location
Chr. 9: 110243812 - 110256640 on Build GRCh38
UniGene ID
Hs.435136
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001244938.1 NP_001231867.1 3 349
NM_003329.3 NP_003320.2 4 409
GenBank mRNA AF065241.1 - 3 141
AF085844.1 - 4 216
AF276919.1 - 3 204
AF313911.1 - 4 214
AK289508.1 - 4 330
AW160756.1 - 4 409
AY004872.1 - 4 214
BC003377.1 - 4 261 2901084
BC054866.1 - 4 280 6463280
BX647247.1 - 3 250
CR407665.1 - 4 204
J04026.1 - 4 267
JQ313905.1 - 4 204
X77584.1 - 4 284

View Full Product Details