Gene Transcripts

Gene Symbol
Vmn1r47
Entrez Gene ID
113846
Gene Name
vomeronasal 1 receptor 47
Gene Aliases
V1RA7, V1ra4
Chromosome Location
Chr. 6: 90021888 - 90022820 on Build GRCm38
UniGene ID
Mm.377143
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053219.2 NP_444449.2 1 797
GenBank mRNA BC111822.1 - 1 796

View Full Product Details