Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_053254.2 | NP_444484.1 | 8 | 879 | |
GenBank mRNA | AB221587.1 | - | 7 | 740 | |
AF145957.2 | - | 8 | 879 | ||
AK007880.1 | - | 2 | 110 | ||
AK135768.1 | - | 8 | 856 | ||
AK154391.1 | - | 5 | 839 | ||
BC023315.1 | - | 8 | 782 | ||
BC048832.1 | - | 5 | 718 | 4506029 |