Gene Transcripts

Gene Symbol
Asb14
Entrez Gene ID
142687
Gene Name
ankyrin repeat and SOCS box-containing 14
Gene Aliases
Asb-14
Chromosome Location
Chr. 14: 26894591 - 26916100 on Build GRCm38
UniGene ID
Mm.138259
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170748.1 NP_001164219.1 10 1840
GenBank mRNA AF403042.1 - 5 1257
AK164800.1 - 4 903
BC145002.1 - 5 1226

View Full Product Details