Gene Transcripts
- Gene Symbol
- Stard5
- Entrez Gene ID
- 170460
- Gene Name
- StAR-related lipid transfer (START) domain containing 5
- Gene Aliases
- 2310058G22Rik, AI607880, D7Ertd152e
- Chromosome Location
- Chr. 7: 83632017 - 83642328 on Build GRCm38
- UniGene ID
- Mm.357953
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_023377.4 |
NP_075866.2
|
1, 2 |
129 |
|
GenBank mRNA |
AF480302.1 |
- |
1, 2 |
105 |
|
AK009980.1 |
- |
1, 2 |
135 |
|
AK077328.1 |
- |
1, 2 |
147 |
|
AK080060.1 |
- |
1, 2 |
147 |
|
AK085852.1 |
- |
1, 2 |
142 |
|
AK142503.1 |
- |
1, 2 |
147 |
|
AK158178.1 |
- |
1, 2 |
131 |
|
AY007808.1 |
- |
1, 2 |
124 |
|
BC031733.1 |
- |
1, 2 |
133 |
|
BC058226.1 |
- |
1, 2 |
129 |
6308417 |
View Full Product Details