Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002590.3 | NP_002581.2 | 3 | 3268 | |
NM_032949.2 | NP_116567.1 | 3 | 2977 | ||
GenBank mRNA | AF061573.2 | - | 3 | 3267 | |
AK293747.1 | - | 4 | 2797 | ||
AK297652.1 | - | 4 | 1846 | ||
AY013873.1 | - | 3 | 2773 | ||
BC036025.1 | - | 3 | 3268 | 5278690 |