Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_176850.2 | NP_789820.2 | 2 | 1428 | |
GenBank mRNA | BC003956.1 | - | 1 | 728 | |
BC037661.1 | - | 2 | 1073 | ||
BC046973.1 | - | 2 | 1143 | ||
DV660897.1 | - | 1 | 461 | ||
GU189417.1 | - | 2 | 981 |