Gene Transcripts

Gene Symbol
Shank2
Entrez Gene ID
210274
Gene Name
SH3/ankyrin domain gene 2
Gene Aliases
ProSAP1, mKIAA1022
Chromosome Location
Chr. 7: 143978056 - 144424494 on Build GRCm38
UniGene ID
Mm.483624
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081370.2 NP_001074839.2 11 4332
NM_001113373.2 NP_001106844.2 16 4778
GenBank mRNA AB099695.1 - 16 4417
AK220363.1 - 9 3347

View Full Product Details