Gene Transcripts

Gene Symbol
Athl1
Entrez Gene ID
212974
Gene Name
ATH1, acid trehalase-like 1 (yeast)
Gene Aliases
5730511L01Rik, BC023151
Chromosome Location
Chr. 7: 140941581 - 140947658 on Build GRCm38
UniGene ID
Mm.260139
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145387.4 NP_663362.2 9 1503
GenBank mRNA AK077662.1 - 9 1504
AK085600.1 - 8 1674
AK171694.1 - 8 1413
AK220240.1 - 5 2735
BC023151.1 - 8 1189
BC056953.1 - 9 1465 5717810

View Full Product Details