Gene Transcripts

Gene Symbol
B4galt7
Entrez Gene ID
218271
Gene Name
xylosylprotein beta1,4-galactosyltransferase, polypeptide 7 (galactosyltransferase I)
Gene Aliases
Chromosome Location
Chr. 13: 55599901 - 55610445 on Build GRCm38
UniGene ID
Mm.139825
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146045.1 NP_666157.1 2 380
GenBank mRNA AK050684.1 - 2 327
AK141739.1 - 2 344
AK146855.1 - 2 427
AK171600.1 - 2 396
BC027195.1 - 2 380 4224150
BC055703.1 - 2 595 6403624

View Full Product Details