Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178684.5 | NP_848799.1 | 3 | 164 | |
GenBank mRNA | AK048061.1 | - | 3 | 202 | |
AK086896.1 | - | 3 | 192 | ||
AK143495.1 | - | 2 | 89 | ||
AK168694.1 | - | 3 | 195 | ||
AK169333.1 | - | 3 | 165 | ||
AK205886.1 | - | 1 | 13 | ||
BC047201.1 | - | 3 | 187 | 6396409 |