Gene Transcripts

Gene Symbol
Ubxn7
Entrez Gene ID
224111
Gene Name
UBX domain protein 7
Gene Aliases
A630090P18, AI196514, Ubxd7, mKIAA0794
Chromosome Location
Chr. 16: 32332184 - 32393750 on Build GRCm38
UniGene ID
Mm.31275
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177633.4 NP_808301.3 11 1442
GenBank mRNA AK129218.1 - 7 1040
AK147730.1 - 11 1438
BC062904.1 - 10 1360

View Full Product Details