Gene Transcripts

Gene Symbol
Ankrd29
Entrez Gene ID
225187
Gene Name
ankyrin repeat domain 29
Gene Aliases
G630054C21, G630054C21Rik, Gm95
Chromosome Location
Chr. 18: 12252357 - 12305720 on Build GRCm38
UniGene ID
Mm.53865
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190371.1 NP_001177300.1 9 855
GenBank mRNA AK090336.1 - 9 808
AK162301.1 - 4 283

View Full Product Details