Gene Transcripts

Gene Symbol
Haus1
Entrez Gene ID
225745
Gene Name
HAUS augmin-like complex, subunit 1
Gene Aliases
BC024400, Ccdc5, HEI-C
Chromosome Location
Chr. 18: 77757567 - 77773847 on Build GRCm38
UniGene ID
Mm.39293
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146089.2 NP_666201.1 2 188
GenBank mRNA AK076912.1 - 2 168
AK146923.1 - 2 185
AK147599.1 - 2 195
BC024400.1 - 2 188 5026575

View Full Product Details