Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145832.3 | NP_665831.1 | 3 | 658 | |
NM_211358.2 | NP_997597.1 | 2 | 1146 | ||
GenBank mRNA | AJ440720.1 | - | 2 | 543 | |
AK030977.1 | - | 3 | 659 | ||
AK033295.1 | - | 2 | 235 | ||
AK040986.1 | - | 2 | 893 | ||
AK045972.1 | - | 3 | 635 | ||
AK050311.1 | - | 3 | 623 | ||
AK081068.1 | - | 2 | 644 | ||
AK088227.1 | - | 3 | 645 | ||
AK138595.1 | - | 2 | 1089 | ||
AK144762.1 | - | 2 | 1068 | ||
AK151154.1 | - | 2 | 1142 | ||
BC031814.1 | - | 2 | 740 | 4196818 | |
BC116736.1 | - | 2 | 571 | 40126055 | |
BC119128.1 | - | 2 | 571 | 8733877 |