Gene Transcripts

Gene Symbol
Amigo1
Entrez Gene ID
229715
Gene Name
adhesion molecule with Ig like domain 1
Gene Aliases
Amigo, ali2
Chromosome Location
Chr. 3: 108186289 - 108192286 on Build GRCm38
UniGene ID
Mm.275752
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004293.2 NP_001004293.1 2 1320
NM_146137.3 NP_666249.2 2 1355
GenBank mRNA AB167509.1 - 1 976
AK035960.1 - 2 1251
AK173118.1 - 2 1309
AY237008.1 - 1 869
BC010598.1 - 2 1271 3498778

View Full Product Details