Gene Transcripts

Gene Symbol
PMAIP1
Entrez Gene ID
5366
Gene Name
phorbol-12-myristate-13-acetate-induced protein 1
Gene Aliases
APR, NOXA
Chromosome Location
Chr. 18: 59899960 - 59904306 on Build GRCh38
UniGene ID
Hs.96
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021127.2 NP_066950.1 2 292
GenBank mRNA AK311943.1 - 2 257
BC013120.1 - 2 215 3858958
BC032663.1 - 3 398 5529754
BF667112.1 - 293
D90070.1 - 2 247
KU178263.1 - 3 225
KU178264.1 - 2 74

View Full Product Details