Gene Transcripts

Gene Symbol
Caprin2
Entrez Gene ID
232560
Gene Name
caprin family member 2
Gene Aliases
C1qdc1, Eeg1, rng140
Chromosome Location
Chr. 6: 148842492 - 148896257 on Build GRCm38
UniGene ID
Mm.150568
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301351.1 NP_001288280.1 6 903
NM_181541.4 NP_853519.2 6 903
GenBank mRNA AK129460.1 - 6 816
AK140744.1 - 6 905
BC025636.1 - 6 860
BC125373.1 - 6 792 40129888
BC125375.1 - 6 792 40129890

View Full Product Details