Gene Transcripts

Gene Symbol
Ano5
Entrez Gene ID
233246
Gene Name
anoctamin 5
Gene Aliases
9330162L24, Gdd1, Tmem16e
Chromosome Location
Chr. 7: 51510875 - 51600693 on Build GRCm38
UniGene ID
Mm.254431
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271879.1 NP_001258808.1 8 976
NM_177694.6 NP_808362.2 9 1018
GenBank mRNA AB125740.1 - 9 1018
AB206762.1 - 9 1018
AK034197.1 - 9 1000
BC109163.2 - 8 898 40047987

View Full Product Details