Gene Transcripts

Gene Symbol
Armc5
Entrez Gene ID
233912
Gene Name
armadillo repeat containing 5
Gene Aliases
AW049683, BC032200
Chromosome Location
Chr. 7: 128237357 - 128245100 on Build GRCm38
UniGene ID
Mm.475117
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146205.2 NP_666317.2 3 1391
GenBank mRNA AK030000.1 - 3 248
AK131111.1 - 3 1391
BC032200.1 - 3 736 5342166
BC089363.1 - 3 632 5720275

View Full Product Details