Gene Transcripts
- Gene Symbol
- POU4F1
- Entrez Gene ID
- 5457
- Gene Name
- POU class 4 homeobox 1
- Gene Aliases
- BRN3A, Oct-T1, RDC-1, brn-3A
- Chromosome Location
- Chr. 13: 78599092 - 78603560 on Build GRCh38
- UniGene ID
- Hs.654522
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006237.3 |
NP_006228.3
|
2 |
1905 |
|
GenBank mRNA |
L20433.1 |
- |
2 |
1908 |
|
- Gene Symbol
- LOC107987188
- Entrez Gene ID
- 107987188
- Gene Name
- uncharacterized LOC107987188
- Gene Aliases
- Chromosome Location
- Chr. 13: 78596140 - 78601130 on Build GRCh38
- Species
- Homo sapiens
View Full Product Details