Gene Transcripts

Gene Symbol
Apol8
Entrez Gene ID
239552
Gene Name
apolipoprotein L 8
Gene Aliases
9830006J20Rik, APOL-II, Apol2
Chromosome Location
Chr. 15: 77745109 - 77757044 on Build GRCm38
UniGene ID
Mm.125650
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081970.1 NP_001075439.1 4 719
GenBank mRNA AK036408.1 - 4 739
AK046043.1 - 4 848
BC129972.1 - 4 719 40090463

View Full Product Details