Gene Transcripts

Gene Symbol
Adnp2
Entrez Gene ID
240442
Gene Name
ADNP homeobox 2
Gene Aliases
8430420L05Rik, BC024969, Kiaa0863, Zfp508, mKIAA0863
Chromosome Location
Chr. 18: 80126315 - 80152063 on Build GRCm38
UniGene ID
Mm.26594
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175028.1 NP_778193.1 4 584
GenBank mRNA AB093268.1 - 4 584
BC044898.1 - 4 489
BC047352.1 - 4 489
BC064672.1 - 2 357

View Full Product Details