Gene Transcripts

Gene Symbol
Styk1
Entrez Gene ID
243659
Gene Name
serine/threonine/tyrosine kinase 1
Gene Aliases
9130025L13, AI326477, Nok
Chromosome Location
Chr. 6: 131299133 - 131317009 on Build GRCm38
UniGene ID
Mm.20114
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172891.2 NP_766479.1 3, 4 1309
GenBank mRNA AK033606.1 - 3, 4 1309
AY563053.1 - 3, 4 464
BC150724.1 - 3, 4 517 9087635

View Full Product Details