Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006437.3 | NP_006428.2 | 22 | 2685 | |
XM_011534931.1 | XP_011533233.1 | 21 | 2735 | ||
XM_011534932.2 | XP_011533234.1 | 22 | 2902 | ||
GenBank mRNA | AF057160.1 | - | 22 | 2631 | |
AF158255.1 | - | 22 | 2685 | ||
AK304399.1 | - | 22 | 2653 | ||
D79999.2 | - | 21 | 2584 |