Gene Transcripts
- Gene Symbol
- Rttn
- Entrez Gene ID
- 246102
- Gene Name
- rotatin
- Gene Aliases
- 4921538A15Rik, AI666264, C530033I08Rik, D230040K24
- Chromosome Location
- Chr. 18: 88971738 - 89131016 on Build GRCm38
- UniGene ID
- Mm.772
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_175542.3 |
NP_780751.2
|
22 |
3118 |
|
GenBank mRNA |
AF265232.1 |
- |
22 |
3118 |
|
View Full Product Details