Gene Transcripts

Gene Symbol
Map3k10
Entrez Gene ID
269881
Gene Name
mitogen-activated protein kinase kinase kinase 10
Gene Aliases
BC028668, BC046514, MST, Mlk2
Chromosome Location
Chr. 7: 27656375 - 27674607 on Build GRCm38
UniGene ID
Mm.389883
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290528.1 NP_001277457.1 1 895
GenBank mRNA AK053830.1 - 1 895

View Full Product Details