Gene Transcripts

Gene Symbol
Slc35e1
Entrez Gene ID
270066
Gene Name
solute carrier family 35, member E1
Gene Aliases
6030458H05, AA408278
Chromosome Location
Chr. 8: 72477995 - 72492614 on Build GRCm38
UniGene ID
Mm.291461
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177766.3 NP_808434.2 6 1168
GenBank mRNA AK031600.1 - 6 916
BC115553.1 - 6 827 40101801
BC115554.1 - 6 827 40101802

View Full Product Details