Gene Transcripts

Gene Symbol
D11Wsu47e
Entrez Gene ID
276852
Gene Name
DNA segment, Chr 11, Wayne State University 47, expressed
Gene Aliases
Chromosome Location
Chr. 11: 113684400 - 113727444 on Build GRCm38
UniGene ID
Mm.239566
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177777.5 NP_808445.2 2 556
GenBank mRNA AK017508.1 - 2 422
AK139389.1 - 3 512
AK171307.1 - 2 465
AK210886.1 - 1 74
BC026908.1 - 1 169 3988907
BC048942.1 - 2 559 6442008

View Full Product Details