Gene Transcripts

Gene Symbol
C2cd3
Entrez Gene ID
277939
Gene Name
C2 calcium-dependent domain containing 3
Gene Aliases
AU020772
Chromosome Location
Chr. 7: 100372233 - 100470159 on Build GRCm38
UniGene ID
Mm.276374
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017985.2 NP_001017985.2 5 1009
GenBank mRNA AK148365.1 - 5 885
AK162155.1 - 5 942
AK165331.1 - 5 903
AK171263.1 - 6 1196
BC094430.1 - 4 590 30643564

View Full Product Details