Gene Transcripts

Gene Symbol
Smo
Entrez Gene ID
319757
Gene Name
smoothened homolog (Drosophila)
Gene Aliases
E130215L21Rik, Smoh, bnb, smoothened
Chromosome Location
Chr. 6: 29735497 - 29761366 on Build GRCm38
UniGene ID
Mm.29279
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176996.4 NP_795970.3 3 1187
GenBank mRNA BC048091.1 - 3 993
BC096028.1 - 3 706 6850065
CN665198.1 - 2 295
FJ883552.1 - 3 696

View Full Product Details