Gene Transcripts

Gene Symbol
Bhlha9
Entrez Gene ID
320522
Gene Name
basic helix-loop-helix family, member a9
Gene Aliases
A830053O21Rik, Fingerin
Chromosome Location
Chr. 11: 76672470 - 76673676 on Build GRCm38
UniGene ID
Mm.131585
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177182.4 NP_796156.3 1 853
GenBank mRNA AK043927.1 - 1 598
BC048728.1 - 1 853 6510150

View Full Product Details