Gene Transcripts

Gene Symbol
Atg2a
Entrez Gene ID
329015
Gene Name
autophagy related 2A
Gene Aliases
1810013C15Rik, A830054M12, BC023754, mKIAA0404
Chromosome Location
Chr. 19: 6241666 - 6262304 on Build GRCm38
UniGene ID
Mm.277384
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_194348.3 NP_919329.2 39 5575
GenBank mRNA AK129132.1 - 39 5500
AK151782.1 - 3 365
BC022657.1 - 2 118
BC023754.1 - 35 4862
BC056482.1 - 25 3142
BC063264.1 - 39 5516 6409527

View Full Product Details