Gene Transcripts

Gene Symbol
Thoc2
Entrez Gene ID
331401
Gene Name
THO complex 2
Gene Aliases
6330441O12Rik, D130005M13Rik, Gm1139, Gm1793, Tho2
Chromosome Location
Chr. X: 41794992 - 41911924 on Build GRCm38
UniGene ID
Mm.259498
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033422.1 NP_001028594.1 32 4265
GenBank mRNA AK076348.1 - 2 68
CO045267.1 - 2 272

View Full Product Details