Gene Transcripts

Gene Symbol
Urb2
Entrez Gene ID
382038
Gene Name
URB2 ribosome biogenesis 2 homolog (S. cerevisiae)
Gene Aliases
C77575, mKIAA0133
Chromosome Location
Chr. 8: 124021442 - 124048505 on Build GRCm38
UniGene ID
Mm.334891
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029876.1 NP_001025047.1 2 220
GenBank mRNA AK133906.1 - 3 534
BC151125.1 - 2 299 9088038

View Full Product Details