Gene Transcripts

Gene Symbol
Slc22a14
Entrez Gene ID
382113
Gene Name
solute carrier family 22 (organic cation transporter), member 14
Gene Aliases
Gm1128
Chromosome Location
Chr. 9: 119169453 - 119190430 on Build GRCm38
UniGene ID
Mm.335525
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037749.2 NP_001032838.1 12 1888
GenBank mRNA BC100471.1 - 12 1888

View Full Product Details