Gene Transcripts

Gene Symbol
Tmem29
Entrez Gene ID
382245
Gene Name
transmembrane protein 29
Gene Aliases
2700081K05Rik, 6330540D07Rik, CXorf44-like
Chromosome Location
Chr. X: 150397773 - 150467331 on Build GRCm38
UniGene ID
Mm.277309
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164686.1 NP_001158158.1 3 330
GenBank mRNA AK012546.1 - 3 341
BC058428.1 - 3 291
CA526769.1 - 3 331
CK624690.1 - 3 301

View Full Product Details