Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001206691.1 | NP_001193620.1 | 10 | 1125 | |
NM_032491.5 | NP_115880.2 | 6 | 791 | ||
NM_213594.2 | NP_998759.1 | 10 | 1372 | ||
GenBank mRNA | AB044245.1 | - | 10 | 1090 | |
AB095365.1 | - | 10 | 1120 | ||
AB095366.1 | - | 10 | 1372 | ||
AF332192.1 | - | 6 | 785 | ||
AK074913.1 | - | 7 | 703 | ||
AK291445.1 | - | 10 | 1357 | ||
AK302837.1 | - | 10 | 1065 | ||
AK315698.1 | - | 6 | 771 | ||
AL833921.1 | - | 3 | 195 | ||
AY102009.1 | - | 10 | 1025 | ||
BC028582.2 | - | 6 | 794 | 4836964 | |
BC030644.1 | - | 6 | 766 | 4825079 |