Gene Transcripts

Gene Symbol
Taar8c
Entrez Gene ID
494546
Gene Name
trace amine-associated receptor 8C
Gene Aliases
taR-8c
Chromosome Location
Chr. 10: 24100843 - 24101951 on Build GRCm38
UniGene ID
Mm.379739
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010840.2 NP_001010840.1 1 633
GenBank mRNA BC139121.1 - 1 633
BC139122.1 - 1 633 8862144

View Full Product Details