Gene Transcripts
- Gene Symbol
- Trav16d-dv11
- Entrez Gene ID
- 547329
- Gene Name
- T cell receptor alpha variable 16D-DV11
- Gene Aliases
- ENSMUSG00000072535, ENSMUSG00000076796, Gm10893, Trav16d/dv11
- Chromosome Location
- Chr. 14: 53047287 - 53047821 on Build GRCm38
- UniGene ID
- Mm.321046
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK088475.1 |
- |
|
233 |
|
U07873.1 |
- |
2 |
187 |
|
View Full Product Details