Gene Transcripts
- Gene Symbol
- Tcrg-V3
- Entrez Gene ID
- 21637
- Gene Name
- T cell receptor gamma, variable 3
- Gene Aliases
- TCRGV5S3, TRGV3, vgamma3
- Chromosome Location
- Chr. 13: 19242845 - 19243301 on Build GRCm38
- UniGene ID
- Mm.364155
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
Z49051.1 |
- |
1 |
200 |
|
- Gene Symbol
- Trgv2
- Entrez Gene ID
- 21636
- Gene Name
- T cell receptor gamma variable 2
- Gene Aliases
- Tcrg-V2, Vgamma2
- Chromosome Location
- Chr. 13: 19336573 - 19337026 on Build GRCm38
- UniGene ID
- Mm.211738
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
Z48589.1 |
- |
1 |
200 |
|
View Full Product Details