Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001026.4 | NP_001017.1 | 3 | 260 | |
NM_001142282.1 | NP_001135754.1 | 3 | 260 | ||
NM_001142283.1 | NP_001135755.1 | 3 | 260 | ||
NM_001142284.1 | NP_001135756.1 | 3 | 260 | ||
NM_001142285.1 | NP_001135757.1 | 3 | 260 | ||
NM_033022.3 | NP_148982.1 | 3 | 260 | ||
GenBank mRNA | AF147330.1 | - | 3 | 141 | |
AK311776.1 | - | 3 | 157 | ||
AK311910.1 | - | 3 | 157 | ||
BC000523.2 | - | 3 | 137 | 2961125 | |
BC003149.1 | - | 3 | 133 | ||
BC071926.1 | - | 3 | 142 | 5556377 | |
BG610671.1 | - | 204 | |||
BG944046.1 | - | 3 | 261 | ||
BM745909.1 | - | 3 | 157 | ||
BX537975.1 | - | 3 | 156 | ||
BX647260.1 | - | 3 | 146 | ||
CD176901.1 | - | 3 | 183 | ||
CD677556.1 | - | 3 | 136 | ||
DB116225.1 | - | 3 | 157 | ||
M31520.1 | - | 3 | 260 |