Gene Transcripts

Gene Symbol
RYR2
Entrez Gene ID
6262
Gene Name
ryanodine receptor 2
Gene Aliases
ARVC2, ARVD2, RYR-2, RyR, VTSIP
Chromosome Location
Chr. 1: 237042208 - 237833988 on Build GRCh38
UniGene ID
Hs.109514
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001035.2 NP_001026.2 14 1332
XM_006711802.3 XP_006711865.1 14 1526
XM_006711803.3 XP_006711866.1 14 1526
XM_006711804.3 XP_006711867.1 14 1526
XM_006711805.3 XP_006711868.1 14 1526
XM_006711806.3 XP_006711869.1 14 1526
XM_006711807.3 XP_006711870.1 14 1526
XM_006711808.3 XP_006711871.1 14 1526
XM_006711809.3 XP_006711872.1 14 1526
XM_006711810.3 XP_006711873.1 14 1526
XM_017002028.1 XP_016857517.1 13 1505
GenBank mRNA X98330.1 - 14 1333

View Full Product Details