Gene Transcripts

Gene Symbol
Prl2c1
Entrez Gene ID
666317
Gene Name
Prolactin family 2, subfamily c, member 1
Gene Aliases
Chromosome Location
Chr. 13: 27849327 - 27857791 on Build GRCm38
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001045532.2 NP_001038997.1 4 440

View Full Product Details