Gene Transcripts

Gene Symbol
SDHD
Entrez Gene ID
6392
Gene Name
succinate dehydrogenase complex subunit D
Gene Aliases
CBT1, CII-4, CWS3, PGL, PGL1, QPs3, SDH4, cybS
Chromosome Location
Chr. 11: 112086824 - 112095801 on Build GRCh38
UniGene ID
Hs.356270
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276503.1 NP_001263432.1 3 274
NM_001276504.1 NP_001263433.1 3 302
NM_001276506.1 NP_001263435.1 5 549
NM_003002.3 NP_002993.1 4 419
NR_077060.1 5 473
GenBank mRNA AB006202.1 - 4 346
AK075360.1 - 4 345
BC005263.1 - 4 363 3930691
BC009574.1 - 4 344 4063808
BC012603.1 - 4 348 4108104
BC015188.1 - 4 370 4279214
BC015992.1 - 4 366 4292983
BC022350.1 - 4 358 4275516
BC070307.1 - 4 344 4277540
BC071755.1 - 4 362 4774525
BC071756.1 - 4 344 6450992
BE567994.1 - 5 366
BF211358.1 - 3 227
BQ932668.1 - 4 427
BT007238.1 - 4 335
CD105572.1 - 3 214
CR456932.1 - 4 335
KU178325.1 - 4 335
KU178326.1 - 3 190

View Full Product Details