Gene Transcripts

Gene Symbol
Ncam1
Entrez Gene ID
24586
Gene Name
neural cell adhesion molecule 1
Gene Aliases
Cd56, N-CAM, N-CAM-1, NCAM-1, NCAM-C, NCAMC, Ncam
Chromosome Location
Chr. 8: 53836797 - 54134881 on Build Rnor_6.0
UniGene ID
Rn.11283
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031521.1 NP_113709.1 5 710
XM_006242980.2 XP_006243042.1 5 861
XM_006242986.2 XP_006243048.1 5 861
XM_006242988.2 XP_006243050.1 5 861
XM_006242994.2 XP_006243056.1 5 861
XM_008766192.1 XP_008764414.1 5 861
XM_008766193.1 XP_008764415.1 5 861
GenBank mRNA BC101924.1 - 5 711
X06564.1 - 5 710

View Full Product Details