Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000341.3 | NP_000332.2 | 3 | 803 | |
XM_011533047.2 | XP_011531349.1 | 3 | 803 | ||
GenBank mRNA | AB033549.1 | - | 3 | 784 | |
AK091932.1 | - | 4 | 669 | ||
AK223146.1 | - | 3 | 765 | ||
AK289636.1 | - | 3 | 781 | ||
AK309145.1 | - | 3 | 797 | ||
AK313157.1 | - | 3 | 797 | ||
BC022386.1 | - | 3 | 801 | 4556946 | |
BC093624.1 | - | 3 | 762 | 7939469 | |
BC093626.1 | - | 3 | 757 | 7939471 | |
DQ023513.1 | - | 3 | 725 | ||
DQ023515.1 | - | 3 | 725 | ||
DQ023516.1 | - | 3 | 725 | ||
DQ023517.1 | - | 3 | 725 | ||
KU178335.1 | - | 3 | 725 | ||
L11696.1 | - | 3 | 768 | ||
M95548.1 | - | 3 | 769 |