Gene Transcripts
- Gene Symbol
- Rab12
- Entrez Gene ID
- 25530
- Gene Name
- RAB12, member RAS oncogene family
- Gene Aliases
- Chromosome Location
- Chr. 9: 114683985 - 114709613 on Build Rnor_6.0
- UniGene ID
- Rn.11021
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013017.1 |
NP_037149.1
|
|
479 |
|
GenBank mRNA |
M83676.1 |
- |
3 |
274 |
|
View Full Product Details