Gene Transcripts

Gene Symbol
Bok
Entrez Gene ID
29884
Gene Name
BCL2-related ovarian killer
Gene Aliases
Bok-BH3
Chromosome Location
Chr. 9: 100829593 - 100840498 on Build Rnor_6.0
UniGene ID
Rn.44461
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017312.2 NP_059008.1 5 1104
GenBank mRNA BC078871.1 - 5 1104
FQ227820.1 - 4 945

View Full Product Details