Gene Transcripts

Gene Symbol
Tpsab1
Entrez Gene ID
54271
Gene Name
tryptase alpha/beta 1
Gene Aliases
Mcpt7, TPSB1, rMCP-7
Chromosome Location
Chr. 10: 14701253 - 14703668 on Build Rnor_6.0
UniGene ID
Rn.10699
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019322.2 NP_062195.2 3 495
GenBank mRNA BC061983.1 - 3 477
U67910.1 - 3 494

View Full Product Details